Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spon1
Entrez Gene ID
233744
Gene Name
spondin 1, (f-spondin) extracellular matrix protein
Gene Aliases
AI666765, AW455831, BC020531, D330035F22Rik, FSP
Chromosome Location
Chr.7: 113765998 - 114043375 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145584.2 NP_663559.1 14 - 15 2323 94
GenBank mRNA AK146506.1 - 14 - 15 2147 94
AK173021.1 - 11 - 12 3858 94
BC020531.1 - 14 - 15 2193 94
BC030339.1 - 5 - 6 777 94

View Full Product Details