Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aatk
Entrez Gene ID
11302
Gene Name
apoptosis-associated tyrosine kinase
Gene Aliases
AATYK, aatyk1, mKIAA0641
Chromosome Location
Chr.11: 120007311 - 120047182 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198785.1 NP_001185714.1 7 - 8 984 66
NM_001198787.1 NP_001185716.1 7 - 8 756 66
NM_007377.4 NP_031403.2 7 - 8 1267 66
GenBank mRNA AB093253.2 - 7 - 8 1267 66
AB288871.1 - 7 - 8 886 66
AF011908.1 - 7 - 8 723 66
AK036705.1 - 7 - 8 758 66
AK147239.1 - 4 - 5 777 66
AK155100.1 - 7 - 8 758 66
AY236858.1 - 7 - 8 762 66
AY236859.1 - 7 - 8 789 66
BC042837.1 - 7 - 8 1289 66
BC052323.1 - 7 - 8 1289 66
BC080846.1 - 7 - 8 838 66

View Full Product Details