Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxa6
Entrez Gene ID
15403
Gene Name
homeobox A6
Gene Aliases
Hox-1.2
Chromosome Location
Chr.6: 52206365 - 52208624 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010454.1 NP_034584.1 1 - 2 443 60
GenBank mRNA AB221546.1 - 1 - 2 443 60
BC119105.1 - 1 - 2 541 60
BC119107.1 - 1 - 2 541 60

View Full Product Details