Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sharpin
Entrez Gene ID
106025
Gene Name
SHANK-associated RH domain interacting protein
Gene Aliases
0610041B22Rik, AW121341, RBCKL1, SIPL1, cpdm
Chromosome Location
Chr.15: 76347040 - 76351115 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025340.2 NP_079616.2 4 - 5 1076 62
GenBank mRNA AB052763.1 - 4 - 5 1062 62
AK002878.1 - 4 - 5 647 62
AK003578.1 - 4 - 5 656 62
AK165824.1 - 3 - 4 856 62
AK171774.1 - 3 - 4 1260 62
AK178102.1 - 1 - 2 82 62
BC016203.1 - 4 - 5 1017 62
BC055758.1 - 4 - 5 1076 62

View Full Product Details