Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ruvbl2
Entrez Gene ID
20174
Gene Name
RuvB-like protein 2
Gene Aliases
mp47, p47
Chromosome Location
Chr.7: 45421896 - 45435170 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011304.3 NP_035434.1 1 - 2 213 58
GenBank mRNA AB013912.1 - 1 - 2 44 58
AK144156.1 - 1 - 2 35 58
AK159112.1 - 1 - 2 215 58
BC072585.1 - - 712 58
BC115809.1 - 1 - 2 31 58
BC116693.1 - 1 - 2 30 58

View Full Product Details