The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175104.4 | NP_780313.1 | 3 - 4 | 523 | 98 |
GenBank mRNA | AK044469.1 | - | 3 - 4 | 327 | 98 |
AK154214.1 | - | 3 - 4 | 327 | 98 | |
BC057111.1 | - | 3 - 4 | 523 | 98 |