Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Obsl1
Entrez Gene ID
98733
Gene Name
obscurin-like 1
Gene Aliases
AW822216
Chromosome Location
Chr.1: 75485825 - 75506452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178884.5 NP_849215.3 7 - 8 2910 73
GenBank mRNA AK029863.1 - 7 - 8 2699 73
AK048144.1 - 7 - 8 2102 73
AK122340.1 - 1 - 2 144 73
AK162473.1 - 7 - 8 2820 73
BC051643.1 - 5 - 6 1462 73
BC056205.1 - 4 - 5 1084 73

View Full Product Details