The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178884.5 | NP_849215.3 | 8 - 9 | 3184 | 87 |
GenBank mRNA | AK029863.1 | - | 8 - 9 | 2973 | 87 |
AK048144.1 | - | 8 - 9 | 2376 | 87 | |
AK122340.1 | - | 2 - 3 | 418 | 87 | |
BC056205.1 | - | 5 - 6 | 1358 | 87 |