Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sytl5
Entrez Gene ID
236643
Gene Name
synaptotagmin-like 5
Gene Aliases
ENSMUSG00000054453, slp5
Chromosome Location
Chr.X: 9747557 - 9998864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290728.1 NP_001277657.1 10 - 11 1373 70
NM_177704.3 NP_808372.1 11 - 12 1439 70
GenBank mRNA AB098160.1 - 11 - 12 1438 70
AB098161.1 - 10 - 11 1372 70
BC150158.1 - 11 - 12 1442 70
BC150480.1 - 11 - 12 1442 70

View Full Product Details