Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgf7
Entrez Gene ID
14178
Gene Name
fibroblast growth factor 7
Gene Aliases
Kgf
Chromosome Location
Chr.2: 126001108 - 126091185 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008008.4 NP_032034.1 1 - 2 189 80
GenBank mRNA AK037172.1 - 1 - 2 191 80
AK086639.1 - 1 - 2 76 80
BC052847.1 - 1 - 2 74 80

View Full Product Details