The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243072.1 | NP_001230001.1 | 16 - 17 | 2100 | 56 |
NM_001243073.1 | NP_001230002.1 | 16 - 17 | 1940 | 56 | |
NM_009152.4 | NP_033178.2 | 15 - 16 | 2368 | 56 | |
GenBank mRNA | AK140817.1 | - | 16 - 17 | 1941 | 56 |
BC057588.1 | - | 15 - 16 | 2015 | 56 | |
BC090844.1 | - | 16 - 17 | 2100 | 56 | |
D85028.2 | - | 15 - 16 | 2370 | 56 | |
L40484.1 | - | 12 - 13 | 1401 | 56 | |
L41541.1 | - | 15 - 16 | 1719 | 56 |