Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmc7
Entrez Gene ID
209760
Gene Name
transmembrane channel-like gene family 7
Gene Aliases
1700030H01Rik, C230064B05, C630024K23Rik
Chromosome Location
Chr.7: 118535841 - 118584790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172476.4 NP_766064.2 11 - 12 1571 62
GenBank mRNA AK083189.1 - 11 - 12 1574 62
AK132820.1 - 11 - 12 1601 62
AY236499.1 - 11 - 12 1632 62
AY263161.1 - 11 - 12 1600 62
BC105966.1 - 5 - 6 602 62

View Full Product Details