The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013551.2 | NP_038579.2 | 1 - 2 | 220 | 82 |
GenBank mRNA | AK151020.1 | - | 1 - 2 | 186 | 82 |
AK153884.1 | - | 1 - 2 | 153 | 82 | |
AK166734.1 | - | 1 - 2 | 118 | 82 | |
AK167702.1 | - | 1 - 2 | 179 | 82 | |
BC003861.1 | - | 1 - 2 | 180 | 82 | |
BY786671.1 | - | 1 - 2 | 221 | 82 |