The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138667.3 | NP_619608.1 | 2 - 3 | 512 | 107 |
GenBank mRNA | AB093262.1 | - | 1 - 2 | 79 | 107 |
AK147830.1 | - | 2 - 3 | 323 | 107 | |
AY093701.1 | - | 1 - 2 | 172 | 107 | |
BC004813.1 | - | 1 - 2 | 179 | 107 | |
CK391470.1 | - | 3 - 4 | 378 | 107 |