Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pamr1
Entrez Gene ID
210622
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
5930437L24, E430002G05Rik, Ramp
Chromosome Location
Chr.2: 102550021 - 102643041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173749.4 NP_776110.3 7 - 8 1209 76
GenBank mRNA AK088017.1 - 7 - 8 1108 76
BC031841.1 - 7 - 8 1163 76
BC057685.1 - 7 - 8 1163 76

View Full Product Details