Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata7
Entrez Gene ID
104871
Gene Name
spermatogenesis associated 7
Gene Aliases
AI661438, B230306G18Rik, HSD3
Chromosome Location
Chr.12: 98628123 - 98669819 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289573.1 NP_001276502.1 3 - 4 370 130
NM_178914.4 NP_849245.2 3 - 4 370 130
GenBank mRNA AK077500.1 - 3 - 4 281 130
AK153698.1 - 3 - 4 346 130
BC046960.1 - 3 - 4 328 130
BY252962.1 - 3 - 4 369 130

View Full Product Details