The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146193.2 | NP_666305.2 | 8 - 8 | 2149 | 153 |
GenBank mRNA | BC031192.1 | - | 7 - 7 | 1627 | 153 |
BC072618.1 | - | 8 - 8 | 2149 | 153 |