The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053256.2 | NP_444486.1 | 3 - 3 | 525 | 96 |
GenBank mRNA | AF399674.1 | - | 2 - 2 | 437 | 96 |