The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198114.2 | NP_932782.2 | 8 - 9 | 946 | 69 |
GenBank mRNA | AK147384.1 | - | 8 - 9 | 904 | 69 |
AY275376.1 | - | 7 - 8 | 846 | 69 | |
BC112414.1 | - | 7 - 8 | 845 | 69 | |
BC148308.2 | - | 7 - 8 | 846 | 69 |