Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam96b
Entrez Gene ID
68523
Gene Name
family with sequence similarity 96, member B
Gene Aliases
1110019N10Rik
Chromosome Location
Chr.8: 104639839 - 104641728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026753.2 NP_081029.1 4 - 5 436 138
GenBank mRNA AK003830.1 - 4 - 5 439 138
BC055880.1 - 4 - 5 410 138

View Full Product Details