Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sostdc1
Entrez Gene ID
66042
Gene Name
sclerostin domain containing 1
Gene Aliases
0610006G05Rik, Sostl, USAG-1, Wise, ectodin
Chromosome Location
Chr.12: 36314169 - 36318452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025312.3 NP_079588.1 1 - 2 334 82
GenBank mRNA AB059271.1 - 1 - 2 209 82
AK002240.1 - 1 - 2 280 82
AK002396.1 - 1 - 2 278 82
AK007893.1 - 1 - 2 279 82
AK007967.1 - 1 - 2 278 82
AY134666.1 - 1 - 2 267 82
AY255635.1 - 1 - 2 278 82
BC021458.1 - 1 - 2 264 82

View Full Product Details