The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145836.2 | NP_665835.1 | 1 - 1 | 3849 | 157 |
GenBank mRNA | AF525300.1 | - | 1 - 1 | 3846 | 158 |
AK154383.1 | - | 1 - 1 | 3600 | 159 | |
AK160059.1 | - | 1 - 1 | 3805 | 159 | |
AK173282.1 | - | 1 - 1 | 3849 | 157 |