The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011675.2 | NP_035805.2 | 7 - 7 | 1295 | 145 |
GenBank mRNA | AK132100.1 | - | 2 - 2 | 567 | 145 |
AK162288.1 | - | 1 - 1 | 299 | 145 | |
BC025146.1 | - | 7 - 7 | 1262 | 145 |