Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfic
Entrez Gene ID
18029
Gene Name
nuclear factor I/C
Gene Aliases
1110019L22Rik, 1500041O16Rik, AA589446, AI746521, NF1-C
Chromosome Location
Chr.10: 81396191 - 81431167 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008688.3 NP_032714.1 7 - 8 1159 74
NM_026756.2 NP_081032.2 7 - 8 1159 74
GenBank mRNA AF358455.1 - 6 - 7 558 74
AF358456.1 - 6 - 7 558 74
AK087539.1 - 7 - 8 1144 74
AK147496.1 - 7 - 8 1144 74
AK155258.1 - 7 - 8 1161 74
U57635.1 - 7 - 8 1099 74
Y07692.1 - 7 - 8 1118 74
Y07693.1 - 7 - 8 1464 74

View Full Product Details