The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039194.3 | NP_001034283.1 | 2 - 3 | 293 | 86 |
NM_001284300.1 | NP_001271229.1 | 2 - 3 | 293 | 86 | |
NM_153779.2 | NP_722474.2 | 2 - 3 | 286 | 86 | |
NM_178058.4 | NP_835159.1 | 2 - 3 | 293 | 86 | |
GenBank mRNA | AK017403.1 | - | 2 - 3 | 291 | 86 |
AK085656.1 | - | 2 - 3 | 245 | 86 | |
AK147741.1 | - | 2 - 3 | 270 | 86 | |
AK155240.1 | - | 2 - 3 | 233 | 86 | |
BC038129.1 | - | 2 - 3 | 286 | 86 |