The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028906.2 | NP_083182.2 | 5 - 6 | 862 | 118 |
GenBank mRNA | AK016546.1 | - | 5 - 6 | 901 | 118 |
AK155605.1 | - | 5 - 6 | 906 | 118 | |
BC043124.1 | - | 5 - 6 | 862 | 118 | |
BC059222.1 | - | 5 - 6 | 862 | 118 |