Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtkn2
Entrez Gene ID
170799
Gene Name
rhotekin 2
Gene Aliases
B130039D23Rik, Mbf, Plekhk1
Chromosome Location
Chr.10: 67979566 - 68043864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081346.1 NP_001074815.1 8 - 9 927 69
GenBank mRNA AK030986.1 - 7 - 8 808 69
AK045134.1 - - 825 69
AK052662.1 - 7 - 8 962 69
BC116341.2 - - 926 69
BC116342.2 - - 926 69
BC158093.1 - 7 - 8 929 69

View Full Product Details