Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fam53c
- Entrez Gene ID
-
66306
- Gene Name
- family with sequence similarity 53, member C
- Gene Aliases
-
2810012G03Rik, Gm21179
- Chromosome Location
-
Chr.18: 34758830 - 34773760 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK135712.1 |
- |
2 - 3 |
273 |
89 |
View Full Product Details