Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tub
- Entrez Gene ID
-
22141
- Gene Name
- tubby candidate gene
- Gene Aliases
-
rd5, tub
- Chromosome Location
-
Chr.7: 109010880 - 109034459 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U52824.1 |
- |
1 - 2 |
241 |
95 |
View Full Product Details