Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrig3
Entrez Gene ID
320398
Gene Name
leucine-rich repeats and immunoglobulin-like domains 3
Gene Aliases
9030421L11Rik, 9130004I02Rik, 9430095K15Rik, mKIAA3016
Chromosome Location
Chr.10: 125966219 - 126015359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177152.5 NP_796126.4 2 - 3 564 75
GenBank mRNA AK137007.1 - 2 - 3 551 75
AY505341.1 - 2 - 3 507 75
BC065142.1 - 2 - 3 530 75

View Full Product Details