The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198091.2 | NP_932759.1 | 1 - 2 | 411 | 67 |
GenBank mRNA | AY255637.1 | - | 1 - 2 | 380 | 67 |