The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177372.3 | NP_796346.2 | 1 - 2 | 141 | 80 |
GenBank mRNA | AK028381.1 | - | 1 - 2 | 118 | 80 |
AK129051.1 | - | 1 - 2 | 126 | 80 | |
BC030382.1 | - | 1 - 2 | 83 | 80 | |
BC115716.1 | - | 1 - 2 | 95 | 80 | |
BY768981.1 | - | 1 - 2 | 143 | 80 |