Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dna2
Entrez Gene ID
327762
Gene Name
DNA replication helicase 2 homolog (yeast)
Gene Aliases
Dna2l, E130315B21Rik
Chromosome Location
Chr.10: 62946980 - 62974188 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177372.3 NP_796346.2 5 - 6 789 113
GenBank mRNA AK028381.1 - 5 - 6 766 113
AK129051.1 - 5 - 6 774 113
BC030382.1 - 5 - 6 731 113
BC115716.1 - 5 - 6 743 113
CO433698.1 - 3 - 4 360 113

View Full Product Details