Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Neurog3
Entrez Gene ID
11925
Gene Name
neurogenin 3
Gene Aliases
Atoh5, Math4B, bHLHa7, ngn3
Chromosome Location
Chr.10: 62133090 - 62134763 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC104326.2 - 1 - 1 295 103
BC104327.2 - 1 - 1 295 103

View Full Product Details