Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gng7
Entrez Gene ID
14708
Gene Name
guanine nucleotide binding protein (G protein), gamma 7
Gene Aliases
AI840417
Chromosome Location
Chr.10: 80948624 - 81014961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010319.3 NP_034449.2 1 - 2 67 62
GenBank mRNA AK141541.1 - 1 - 2 39 62
AK147843.1 - 1 - 2 30 62
AY035844.1 - 1 - 2 50 62
BY271138.1 - 1 - 2 87 62

View Full Product Details