Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpf1
Entrez Gene ID
29861
Gene Name
D4, zinc and double PHD fingers family 1
Gene Aliases
Neud4
Chromosome Location
Chr.7: 29303959 - 29317592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013874.2 NP_038902.1 7 - 8 804 64
GenBank mRNA AK142419.1 - 7 - 8 769 64
BC052348.1 - 7 - 8 833 64
U48238.1 - 7 - 8 803 64

View Full Product Details