Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clip3
Entrez Gene ID
76686
Gene Name
CAP-GLY domain containing linker protein 3
Gene Aliases
1500005P14Rik, AI844915, Clipr-59
Chromosome Location
Chr.7: 30291688 - 30308367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081114.1 NP_001074583.1 8 - 9 1204 68
GenBank mRNA BC138413.1 - 7 - 8 1123 68

View Full Product Details