Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankk1
Entrez Gene ID
244859
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
9930020N01Rik
Chromosome Location
Chr.9: 49412955 - 49427791 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172922.3 NP_766510.1 6 - 7 1020 74
GenBank mRNA AK036872.1 - 6 - 7 1021 74
BC145078.1 - 6 - 7 1010 74

View Full Product Details