Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tube1
Entrez Gene ID
71924
Gene Name
epsilon-tubulin 1
Gene Aliases
2310061K05Rik, AI551343, Tube
Chromosome Location
Chr.10: 39133979 - 39151058 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028006.2 NP_082282.1 3 - 4 215 97
GenBank mRNA AK010005.1 - 3 - 4 209 97
AK039886.1 - 3 - 4 212 97
AK041416.1 - 3 - 4 180 97
AK087403.1 - 3 - 4 170 97
AK148039.1 - 3 - 4 215 97
BC062179.1 - 3 - 4 212 97

View Full Product Details