The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285885.1 | NP_001272814.1 | 1 - 2 | 1158 | 98 |
NM_001285887.1 | NP_001272816.1 | 2 - 3 | 351 | 98 | |
NM_001285888.1 | NP_001272817.1 | 2 - 3 | 351 | 98 | |
NM_177462.5 | NP_803413.1 | 2 - 3 | 351 | 98 | |
NR_104366.1 | 1 - 2 | 1158 | 98 | ||
GenBank mRNA | AK035136.1 | - | 2 - 3 | 353 | 98 |
AK081836.1 | - | 2 - 3 | 341 | 98 | |
AK082594.1 | - | 1 - 2 | 1100 | 98 | |
AK170695.1 | - | 1 - 2 | 1120 | 98 | |
BC094598.1 | - | 1 - 2 | 1131 | 98 | |
BC140958.1 | - | 2 - 3 | 226 | 98 | |
BC144807.1 | - | 2 - 3 | 226 | 98 |