The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013622.3 | NP_038650.3 | 2 - 3 | 682 | 111 |
GenBank mRNA | AK043873.1 | - | 2 - 3 | 683 | 111 |
BC137958.1 | - | 2 - 3 | 609 | 111 | |
BC137969.1 | - | 2 - 3 | 609 | 111 | |
L06322.1 | - | 2 - 3 | 639 | 111 | |
L07271.1 | - | 2 - 3 | 609 | 111 | |
L11064.1 | - | 2 - 3 | 592 | 111 | |
S65335.1 | - | 2 - 3 | 608 | 111 | |
S66181.1 | - | 2 - 3 | 560 | 111 |