Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a16
Entrez Gene ID
70840
Gene Name
solute carrier family 22 (organic cation transporter), member 16
Gene Aliases
4921504E14Rik, CT2, FLIPT2, OCT6, OKB1
Chromosome Location
Chr.10: 40570321 - 40604135 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027572.1 NP_081848.1 1 - 2 157 86
GenBank mRNA BC100473.1 - 1 - 2 157 86
BC138326.1 - 1 - 2 168 86
BC138327.1 - 1 - 2 168 86
DQ164804.2 - 1 - 2 183 86

View Full Product Details