The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027572.1 | NP_081848.1 | 7 - 8 | 1626 | 78 |
GenBank mRNA | AK014809.1 | - | 4 - 5 | 859 | 78 |
BC100473.1 | - | 7 - 8 | 1626 | 78 | |
BC138326.1 | - | 7 - 8 | 1637 | 78 | |
BC138327.1 | - | 7 - 8 | 1637 | 78 | |
DQ164804.2 | - | 7 - 8 | 1652 | 78 | |
DQ164805.1 | - | 7 - 8 | 1613 | 78 |