Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Maneal
Entrez Gene ID
215090
Gene Name
mannosidase, endo-alpha-like
Gene Aliases
Gm50
Chromosome Location
Chr.4: 124855239 - 124880938 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007573.2 NP_001007574.1 3 - 4 728 94
GenBank mRNA BC065047.1 - - 747 94

View Full Product Details