The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026031.3 | NP_080307.1 | 7 - 8 | 730 | 76 |
GenBank mRNA | AK007361.1 | - | 5 - 6 | 501 | 76 |
AK008801.1 | - | 3 - 4 | 282 | 76 | |
AK012550.1 | - | 7 - 8 | 731 | 76 | |
AK145266.1 | - | 7 - 8 | 763 | 76 | |
AK147113.1 | - | 7 - 8 | 761 | 76 | |
BC043139.1 | - | 5 - 6 | 3819 | 76 | |
BC061074.1 | - | 7 - 8 | 759 | 76 |