The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207210.2 | NP_997093.2 | 14 - 15 | 1723 | 70 |
GenBank mRNA | AK077117.1 | - | 14 - 15 | 1724 | 70 |
BC052324.1 | - | 13 - 14 | 1768 | 70 |