Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mug1
Entrez Gene ID
17836
Gene Name
murinoglobulin 1
Gene Aliases
-
Chromosome Location
Chr.6: 121838497 - 121889057 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008645.3 NP_032671.2 27 - 28 3455 58
GenBank mRNA AK144255.1 - 27 - 28 3419 58
AK144541.1 - 27 - 28 3413 58
AK149376.1 - 26 - 27 3678 58
BC036299.1 - 27 - 28 3401 58
BC049868.1 - 27 - 28 3451 58
BC051037.1 - 27 - 28 3455 58
M65736.1 - 27 - 28 3396 58

View Full Product Details