Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubxn11
Entrez Gene ID
67586
Gene Name
UBX domain protein 11
Gene Aliases
4930506L07Rik, D4Bwg1540e, Soc, Soci, Ubxd5
Chromosome Location
Chr.4: 134102512 - 134127621 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026257.3 NP_080533.1 13 - 14 1242 63
NR_045571.1 12 - 13 1121 63
GenBank mRNA AK010492.1 - 6 - 7 721 63
AK015720.1 - 13 - 14 1226 63
AK048494.1 - 13 - 14 1244 63

View Full Product Details