Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ect2l
- Entrez Gene ID
-
100045792
- Gene Name
- epithelial cell transforming sequence 2 oncogene-like
- Gene Aliases
-
C330021H03Rik, ENSMUSG00000071392, Gm10331
- Chromosome Location
-
Chr.10: 18129022 - 18210890 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK021214.1 |
- |
3 - 4 |
380 |
95 |
View Full Product Details