The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153402.2 | NP_700451.2 | 2 - 3 | 441 | 110 |
GenBank mRNA | AB081473.1 | - | 2 - 3 | 293 | 110 |
AK035042.1 | - | 2 - 3 | 407 | 110 | |
AK042049.1 | - | 2 - 3 | 363 | 110 | |
AK083652.1 | - | 2 - 3 | 408 | 110 | |
AK154436.1 | - | 2 - 3 | 442 | 110 | |
AK171118.1 | - | 2 - 3 | 442 | 110 | |
AK172226.1 | - | 2 - 3 | 363 | 110 | |
BC137964.1 | - | 2 - 3 | 288 | 110 | |
BC145061.1 | - | 2 - 3 | 288 | 110 | |
BC145063.1 | - | 2 - 3 | 288 | 110 |